3-(2,2-Dimethylpropoxy)phenol 95%

Lieferant: Apollo Scientific

OR957636-1G
APOSOR957636-1GEA 632 EUR
APOSOR957636-1G
3-(2,2-Dimethylpropoxy)phenol 95%
3-(2,2-Dimethylpropoxy)phenol
Formel: C₁₁H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23111162
CAS-Nummer: 1394975-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR