2-[(tert-Butoxy)methyl]phenol 95%

Lieferant: Apollo Scientific

OR957637-1G
APOSOR957637-1GEA 632 EUR
APOSOR957637-1G
2-[(tert-Butoxy)methyl]phenol 95%
2-[(tert-Butoxy)methyl]phenol
Formel: C₁₁H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD24716028
CAS-Nummer: 225651-31-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR