3-[(tert-Butoxy)methyl]phenol 98%

Lieferant: Apollo Scientific

OR957638-1G
APOSOR957638-1GEA 632 EUR
APOSOR957638-1G
3-[(tert-Butoxy)methyl]phenol 98%
3-[(tert-Butoxy)methyl]phenol
Formel: C₁₁H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD24726486
CAS-Nummer: 402592-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR