4-[(tert-Butoxy)methyl]phenol 98%

Lieferant: Apollo Scientific

OR957639-1G
APOSOR957639-1GEA 632 EUR
APOSOR957639-1G
4-[(tert-Butoxy)methyl]phenol 98%
4-[(tert-Butoxy)methyl]phenol
Formel: C₁₁H₁₆O₂
Molecular Weight: 180,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23111241
CAS-Nummer: 282713-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR