4-Chloro-2-(propan-2-yloxy)phenol 98%

Lieferant: Apollo Scientific

OR957640-1G
APOSOR957640-1GEA 364 EUR
APOSOR957640-1G
4-Chloro-2-(propan-2-yloxy)phenol 98%
4-Chloro-2-(propan-2-yloxy)phenol
Formel: C₉H₁₁ClO₂
Molecular Weight: 186,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997335
CAS-Nummer: 18113-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR