4-Chloro-3-isopropoxyphenol 95%

Lieferant: Apollo Scientific

OR957641-1G
APOSOR957641-1GEA 632 EUR
APOSOR957641-1G
4-Chloro-3-isopropoxyphenol 95%
4-Chloro-3-isopropoxyphenol
Formel: C₉H₁₁ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16997333
CAS-Nummer: 1243454-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR