Ethyl 3-(3-hydroxyphenyl)propanoate 95%

Lieferant: Apollo Scientific

OR957643-1G
APOSOR957643-1GEA 364 EUR
APOSOR957643-1G
Ethyl 3-(3-hydroxyphenyl)propanoate 95%
Ethyl 3-(3-hydroxyphenyl)propanoate
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772960
CAS-Nummer: 34708-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR