Ethyl 2-(3-hydroxyphenoxy)acetate 95%
Lieferant: Apollo Scientific
OR957645-1G
APOSOR957645-1GEA
632
EUR
APOSOR957645-1G
Ethyl 2-(3-hydroxyphenoxy)acetate 95%
Ethyl 2-(3-hydroxyphenoxy)acetate
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23111306 CAS-Nummer: 68747-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...