4,5-Dichloro-2-methoxyphenol 98%

Lieferant: Apollo Scientific

OR957647-1G
APOSOR957647-1GEA 364 EUR
APOSOR957647-1G
4,5-Dichloro-2-methoxyphenol 98%
4,5-Dichloro-2-methoxyphenol
Formel: C₇H₆Cl₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01707578
CAS-Nummer: 2460-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR