4-Methoxy-3-(propan-2-yloxy)phenol 95%

Lieferant: Apollo Scientific

OR957649-1G
APOSOR957649-1GEA 632 EUR
APOSOR957649-1G
4-Methoxy-3-(propan-2-yloxy)phenol 95%
4-Methoxy-3-(propan-2-yloxy)phenol
Formel: C₁₀H₁₄O₃
Molecular Weight: 182.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16998773
CAS-Nummer: 86636-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR