3-Chloro-4,5-dimethoxyphenol 95%
Lieferant: Apollo Scientific
OR957650-1G
APOSOR957650-1GEA
632
EUR
APOSOR957650-1G
3-Chloro-4,5-dimethoxyphenol 95%
3-Chloro-4,5-dimethoxyphenol
Formel:
C₈H₉ClO₃ Molecular Weight: 188,608 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24717705 CAS-Nummer: 89981-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...