2,3,4-Trimethoxyphenol 95%

Lieferant: Apollo Scientific

OR957651-1G
APOSOR957651-1GEA 104 EUR
APOSOR957651-1G
2,3,4-Trimethoxyphenol 95%
2,3,4-Trimethoxyphenol
Formel: C₉H₁₂O₄
Molecular Weight: 184.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068804
CAS-Nummer: 19676-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR