4'-Chloro-3'-hydroxypropiophenone 95%

Lieferant: Apollo Scientific

OR957664-1G
APOSOR957664-1GEA 632 EUR
APOSOR957664-1G
4'-Chloro-3'-hydroxypropiophenone 95%
4'-Chloro-3'-hydroxypropiophenone
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393261
CAS-Nummer: 1261516-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR