4-(Trimethylsilyl)phenol 95%

Lieferant: Apollo Scientific

OR957666-1G
APOSOR957666-1GEA 632 EUR
APOSOR957666-1G
4-(Trimethylsilyl)phenol 95%
4-(Trimethylsilyl)phenol
Formel: C₉H₁₄OSi
Molecular Weight: 166.292 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00034672
CAS-Nummer: 13132-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR