3-Hydroxy-N-(propan-2-yl)benzamide 95%

Lieferant: Apollo Scientific

OR957669-1G
APOSOR957669-1GEA 632 EUR
APOSOR957669-1G
3-Hydroxy-N-(propan-2-yl)benzamide 95%
3-Hydroxy-N-(propan-2-yl)benzamide
Formel: C₁₀H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11130643
CAS-Nummer: 80917-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR