4-Hydroxy-N-methoxy-N-methylbenzamide 95%

Lieferant: Apollo Scientific

OR957681-1G
APOSOR957681-1GEA 632 EUR
APOSOR957681-1G
4-Hydroxy-N-methoxy-N-methylbenzamide 95%
4-Hydroxy-N-methoxy-N-methylbenzamide
Formel: C₉H₁₁NO₃
Molecular Weight: 181,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09029637
CAS-Nummer: 460747-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR