3-Hydroxy-N-methoxy-N-methylbenzamide 95%
Lieferant: Apollo Scientific
OR957682-1G
APOSOR957682-1GEA
632
EUR
APOSOR957682-1G
3-Hydroxy-N-methoxy-N-methylbenzamide 95%
3-Hydroxy-N-methoxy-N-methylbenzamide
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18843718 CAS-Nummer: 615558-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...