N-(3-Hydroxyphenyl)methanesulphonamide 98%

Lieferant: Apollo Scientific

OR957683-1G
APOSOR957683-1GEA 502 EUR
APOSOR957683-1G
N-(3-Hydroxyphenyl)methanesulphonamide 98%
N-(3-Hydroxyphenyl)methanesulphonamide
Formel: C₇H₉NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD04151187
CAS-Nummer: 71290-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR