N-(4-Hydroxyphenyl)methanesulphonamide 95%

Lieferant: Apollo Scientific

Synonyme: Dofetilide Impurity 15

OR957684-1G
APOSOR957684-1GEA 114 EUR
APOSOR957684-1G
N-(4-Hydroxyphenyl)methanesulphonamide 95%
N-(4-Hydroxyphenyl)methanesulphonamide
Formel: C₇H₉NO₃S
Molecular Weight: 187,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00720883
CAS-Nummer: 51767-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR