N-(4-Chloro-2-hydroxyphenyl)acetamide 95%

Lieferant: Apollo Scientific

OR957685-1G
APOSOR957685-1GEA 458 EUR
APOSOR957685-1G
N-(4-Chloro-2-hydroxyphenyl)acetamide 95%
N-(4-Chloro-2-hydroxyphenyl)acetamide
Formel: C₈H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02293384
CAS-Nummer: 16323-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR