2,6-Dichloro-3-hydroxybenzonitrile 95%

Lieferant: Apollo Scientific

OR957691-1G
APOSOR957691-1GEA 671 EUR
APOSOR957691-1G
2,6-Dichloro-3-hydroxybenzonitrile 95%
2,6-Dichloro-3-hydroxybenzonitrile
Formel: C₇H₃Cl₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD01662155
CAS-Nummer: 3336-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR