3-Chloro-4-(cyclopropylmethoxy)phenol 95%

Lieferant: Apollo Scientific

OR957693-1G
APOSOR957693-1GEA 364 EUR
APOSOR957693-1G
3-Chloro-4-(cyclopropylmethoxy)phenol 95%
3-Chloro-4-(cyclopropylmethoxy)phenol
Formel: C₁₀H₁₁ClO₂
Molecular Weight: 198,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17000220
CAS-Nummer: 1243284-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR