N-Cyclopropyl-4-hydroxybenzamide 95%

Lieferant: Apollo Scientific

OR957694-1G
APOSOR957694-1GEA 451 EUR
APOSOR957694-1G
N-Cyclopropyl-4-hydroxybenzamide 95%
N-Cyclopropyl-4-hydroxybenzamide
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11130694
CAS-Nummer: 860298-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR