1-Hydroxy-2-methoxynaphthalene 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxynaphthalen-1-ol

OR957697-1G
APOSOR957697-1GEA 632 EUR
APOSOR957697-1G
1-Hydroxy-2-methoxynaphthalene 95%
1-Hydroxy-2-methoxynaphthalene
Formel: C₁₁H₁₀O₂
Molecular Weight: 174,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14534802
CAS-Nummer: 1888-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR