4-(5-Methyl-1,3,4-oxadiazol-2-yl)phenol 95%

Lieferant: Apollo Scientific

OR957701-1G
APOSOR957701-1GEA 261 EUR
APOSOR957701-1G
4-(5-Methyl-1,3,4-oxadiazol-2-yl)phenol 95%
4-(5-Methyl-1,3,4-oxadiazol-2-yl)phenol
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04126174
CAS-Nummer: 25877-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR