N,N-Diethyl-4-hydroxybenzamide 95%

Lieferant: Apollo Scientific

OR957704-1G
APOSOR957704-1GEA 538 EUR
APOSOR957704-1G
N,N-Diethyl-4-hydroxybenzamide 95%
N,N-Diethyl-4-hydroxybenzamide
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01120247
CAS-Nummer: 79119-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR