Ethyl 3-hydroxy-4-methoxybenzoate 95%

Lieferant: Apollo Scientific

OR957711-1G
APOSOR957711-1GEA 538 EUR
APOSOR957711-1G
Ethyl 3-hydroxy-4-methoxybenzoate 95%
Ethyl 3-hydroxy-4-methoxybenzoate
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204303
CAS-Nummer: 148527-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR