5-tert-butyl-2-ethoxyphenol 95%

Lieferant: Apollo Scientific

OR957726-1G
APOSOR957726-1GEA 574 EUR
APOSOR957726-1G
5-tert-butyl-2-ethoxyphenol 95%
5-tert-butyl-2-ethoxyphenol
Formel: C₁₂H₁₈O₂
Molecular Weight: 194,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14537713
CAS-Nummer: 1216282-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR