tert-Butyl 3-hydroxybenzoate 96%

Lieferant: Apollo Scientific

OR957743-1G
APOSOR957743-1GEA 207 EUR
APOSOR957743-1G
tert-Butyl 3-hydroxybenzoate 96%
tert-Butyl 3-hydroxybenzoate
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953071
CAS-Nummer: 57704-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR