4-Ethoxy-3-nitrophenol 95%

Lieferant: Apollo Scientific

OR957779-1G
APOSOR957779-1GEA 632 EUR
APOSOR957779-1G
4-Ethoxy-3-nitrophenol 95%
4-Ethoxy-3-nitrophenol
Formel: C₈H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD23111049
CAS-Nummer: 1394969-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR