4-(Dimethylamino)-3-nitrophenol 95%

Lieferant: Apollo Scientific

OR957814-1G
APOSOR957814-1GEA 632 EUR
APOSOR957814-1G
4-(Dimethylamino)-3-nitrophenol 95%
4-(Dimethylamino)-3-nitrophenol
Formel: C₈H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20926279
CAS-Nummer: 14703-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR