trans-3-Hydroxy-β-nitrostyrol 95%
Lieferant: Apollo Scientific
Synonyme:
1-(3-Hydroxyphenyl)-2-nitroethene
OR957830-5G
APOSOR957830-5GEA
487
EUR
APOSOR957830-5G
trans-3-Hydroxy-β-nitrostyrol 95%
trans-3-Hydroxy-β-nitrostyrol
Formel:
C₈H₇NO₃ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
3156-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...