8-Bromoimidazo[1,2-a]pyrazine 97%

Lieferant: Apollo Scientific
Achtung

OR957970-250MG
APOSOR957970-250MGEA 56.7 EUR
APOSOR957970-250MG
8-Bromoimidazo[1,2-a]pyrazine 97%
8-Bromoimidazo[1,2-a]pyrazine
Formel: C₆H₄BrN₃
Lagertemperatur: Kühlschrank
MDL: MFCD18072715
CAS-Nummer: 69214-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR