4-Butyl-2-nitroaniline 95%
Lieferant: Apollo Scientific
OR957993-5G
APOSOR957993-5GEA
121
EUR
APOSOR957993-5G
4-Butyl-2-nitroaniline 95%
4-Butyl-2-nitroaniline
Formel:
C₁₀H₁₄N₂O₂ Molecular Weight: 194.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23725964 CAS-Nummer: 3663-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...