5-Bromo-2-(bromomethyl)pyrimidine 97+%

Lieferant: Apollo Scientific
Achtung

OR957999-250MG
APOSOR957999-250MGEA 82.8 EUR
APOSOR957999-250MG
5-Bromo-2-(bromomethyl)pyrimidine 97+%
5-Bromo-2-(bromomethyl)pyrimidine
Formel: C₅H₄Br₂N₂
Molecular Weight: 251,91 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16606298
CAS-Nummer: 1193116-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR