(8-Methylnaphthalen-1-yl)methanol 95%
Lieferant: Apollo Scientific
OR958017-1G
APOSOR958017-1GEA
647
EUR
APOSOR958017-1G
(8-Methylnaphthalen-1-yl)methanol 95%
(8-Methylnaphthalen-1-yl)methanol
Formel:
C₁₂H₁₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD18072722 CAS-Nummer: 10336-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...