(3R)-3-Phenylpyrrolidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR958065-250MG
APOSOR958065-250MGEA
947
EUR
APOSOR958065-250MG
(3R)-3-Phenylpyrrolidine 95%
(3R)-3-Phenylpyrrolidine
Formel:
C₁₀H₁₃N Molecular Weight: 147,217 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19216485 CAS-Nummer: 61586-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...