2,3-Dichloro-6-hydroxypyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5,6-Dichloropyridin-2-ol, 5,6-Dichloro-2(1H)-pyridinone
OR958310-250MG
APOSOR958310-250MGEA
575
EUR
APOSOR958310-250MG
2,3-Dichloro-6-hydroxypyridine
2,3-Dichloro-6-hydroxypyridine
Formel:
C₅H₃Cl₂NO Molecular Weight: 163,989 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16610775 CAS-Nummer: 24525-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...