8-Chloro-4-methylquinolin-2-ol 95%

Lieferant: Apollo Scientific

Synonyme: 8-Chloro-4-methylquinolin-2(1H)-one

OR958392-100MG OR958392-1G OR958392-250MG
APOSOR958392-100MGEA 58 EUR
APOSOR958392-100MG APOSOR958392-1G APOSOR958392-250MG
8-Chloro-4-methylquinolin-2-ol 95%
8-Chloro-4-methylquinolin-2-ol
Formel: C₁₀H₈ClNO
Molecular Weight: 193.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11868958
CAS-Nummer: 54904-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR