4-Indolcarbonsäure 98%

Lieferant: Apollo Scientific
Achtung

OR9584-25G
APOSOR9584-25GEA 115 EUR
APOSOR9584-25G
4-Indolcarbonsäure 98%
4-Indolcarbonsäure
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Schmelzpunkt: 207…214 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009739
CAS-Nummer: 2124-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR