Ethyl 5-chloro-1-methyl-1H-pyrazole-4-carboxylate 95%

Lieferant: Apollo Scientific

OR958537-250MG
APOSOR958537-250MGEA 21.9 EUR
APOSOR958537-250MG
Ethyl 5-chloro-1-methyl-1H-pyrazole-4-carboxylate 95%
Ethyl 5-chloro-1-methyl-1H-pyrazole-4-carboxylate
Formel: C₇H₉ClN₂O₂
Molecular Weight: 188,612 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265503
CAS-Nummer: 56984-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR