2-Methoxy-6-methylpyridin-4-amine 95%
Lieferant: Apollo Scientific
OR958656-1G
APOSOR958656-1GEA
121
EUR
APOSOR958656-1G
2-Methoxy-6-methylpyridin-4-amine 95%
2-Methoxy-6-methylpyridin-4-amine
Formel:
C₇H₁₀N₂O Molecular Weight: 138.167 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15529703 CAS-Nummer: 89943-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...