5-(4-Pyridyl)-1H-tetrazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1H-Tetrazol-5-yl)pyridine
OR9588-5G
APOSOR9588-5GEA
151
EUR
APOSOR9588-5G
5-(4-Pyridyl)-1H-tetrazol
5-(4-Pyridyl)-1H-tetrazole
Formel:
C₆H₅N₅ Molecular Weight: 147,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068115 CAS-Nummer: 14389-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...