2,3-Dihydrobenzo[b]furan-7-ylamine 95%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dihydrobenzofuran-7-amine

OR958944-1G
APOSOR958944-1GEA 84.1 EUR
APOSOR958944-1G
2,3-Dihydrobenzo[b]furan-7-ylamine 95%
2,3-Dihydrobenzo[b]furan-7-ylamine
Formel: C₈H₉NO
Molecular Weight: 135.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109313
CAS-Nummer: 13414-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR