cis-1,2-Cyclohexanedicarboximide 95%

Lieferant: Apollo Scientific

OR959262-100G
APOSOR959262-100GEA 40.6 EUR
APOSOR959262-100G
cis-1,2-Cyclohexanedicarboximide 95%
cis-1,2-Cyclohexanedicarboximide
Formel: C₈H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00067706
CAS-Nummer: 7506-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR