N-(3-Hydroxypyridin-2-yl)acetamide 96%
Lieferant: Apollo Scientific
OR959383-5G
APOSOR959383-5GEA
75.4
EUR
APOSOR959383-5G
N-(3-Hydroxypyridin-2-yl)acetamide 96%
N-(3-Hydroxypyridin-2-yl)acetamide
Formel:
C₇H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09757484 CAS-Nummer: 31354-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...