2-Methoxypyrimidine-5-carbonitrile 98%

Lieferant: Apollo Scientific

OR959401-1G
APOSOR959401-1GEA 250 EUR
APOSOR959401-1G
2-Methoxypyrimidine-5-carbonitrile 98%
2-Methoxypyrimidine-5-carbonitrile
Formel: C₆H₅N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11840202
CAS-Nummer: 38373-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR