3-Amino-5-bromopyridine-2-carbonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-5-bromopicolinonitrile

OR959411-5G
APOSOR959411-5GEA 150 EUR
APOSOR959411-5G
3-Amino-5-bromopyridine-2-carbonitrile 98%
3-Amino-5-bromopyridine-2-carbonitrile
Formel: C₆H₄BrN₃
Molecular Weight: 198.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375025
CAS-Nummer: 573675-27-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR