3-Chloro-2-(cyclopentyloxy)pyridine 97%

Lieferant: Apollo Scientific

OR959435-1G
APOSOR959435-1GEA 270 EUR
APOSOR959435-1G
3-Chloro-2-(cyclopentyloxy)pyridine 97%
3-Chloro-2-(cyclopentyloxy)pyridine
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21609701
CAS-Nummer: 1289193-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR