3-Chloro-2-(2-methylpropoxy)pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-2-Isobutoxypyridine

OR959437-1G
APOSOR959437-1GEA 304 EUR
APOSOR959437-1G
3-Chloro-2-(2-methylpropoxy)pyridine 95%
3-Chloro-2-(2-methylpropoxy)pyridine
Formel: C₉H₁₂ClNO
Molecular Weight: 185,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21609702
CAS-Nummer: 1288989-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR